Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Mucochloric acid, 99% (dry wt.), water <4.0%
Click to view available options
Quantity:
500g
Specifications
Density | 1.85 |
Melting Point | 125°C to 128°C (dried) |
Quantity | 500g |
Flash Point | 186°C (367°F) |
UN Number | UN2923 |
Beilstein | 1705641 |
Merck Index | 14,6297 |
Formula Weight | 168.96 |
Percent Purity | 99% |
Chemical Name or Material | Mucochloric acid |
Show More |
Safety and Handling
GHS H Statement
H301-H314-H318-H341
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H301-H314-H317-H335-H340
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 201-752-4
RTECSNumber : AS7700000
TSCA : Yes
Recommended Storage : Ambient temperatures