Learn More
Invitrogen™ MUL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109996
Description
Immunogen sequence: MESGGRPSLC QFILLGTTSV VTAALYSVYR QKARVSQELK GAKKVHLGED LKSILSEAPG KCVPYAVIEG AVRSVKETLN SQFVENCKGV IQRLTLQEHK MVWNRTTHLW NDCSKIIHQR TNTVPFDLVP HEDGVDVAVR VLKPLDSVDL GLETVYEKFH PSIQSFTDVI GHYISGERPK GIQETEEMLK VGATLTGVGE LVLDNNSVRL QPPKQGMQYY LSSQDFDSLL QRQESSVRLW KVLALVFGFA TCATLFFILR KQYLQRQERL RLKQMQEEFQ EHEAQLLSRA KPEDRESLKS ACVVCLSSFK SCVFLECGHV CSCTECYRAL PEPKKCPICR QAITRVIPLY NS.
E3 ubiquitin-protein ligase that plays a role in the control of mitochondrial morphology. Promotes mitochondrial fragmentation and influences mitochondrial localization. Inhibits cell growth. When overexpressed, activates JNK through MAP3K7/TAK1 and induces caspase-dependent apoptosis. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.
Specifications
MUL1 | |
Polyclonal | |
Unconjugated | |
MUL1 | |
0610009K11Rik; AV000801; C1orf166; E3 SUMO-protein ligase MUL1; E3 ubiquitin ligase; E3 ubiquitin-protein ligase MUL1; Gide; growth inhibition and death E3 ligase; MAPL; mitochondria-anchored protein ligase; mitochondrial E3 ubiquitin ligase 1; mitochondrial E3 ubiquitin protein ligase 1; mitochondrial ubiquitin ligase activator of NF-kB; mitochondrial ubiquitin ligase activator of NFKB 1; Mitochondrial-anchored protein ligase; MUL1; MULAN; Putative NF-kappa-B-activating protein 266; RGD1309944; ring finger protein 218; RING-type E3 ubiquitin transferase NFKB 1; RNF218; RP11-401M16.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
68350, 79594 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.06 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8VCM5, Q969V5 | |
MUL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-352 of human MUL1 (NP_078820.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.