Learn More
Kamiya Biomedical Multi-Analyte Calibrator, 6x1mL/PK RX
SDP
Supplier:  Kamiya Biomedical KAI016C
Encompass_Preferred
K-ASSAY Multi-Analyte Calibrator. 6 x 1 mL (liquid). For 6 point calibration. The K-ASSAY Multi-Analyte Calibrator is intended to be used for the calibration of the K-ASSAY Alpha-1 Acid Glycoprotein (Cat. No. KAI-021), Alpha-1 Anti-Trypsin (Cat. No. KAI-001), Complement C3 (Cat. No. KAI-009), Complement C4 (Cat. No. KAI-010), Haptoglobin (Cat. No. KAI-022), IgA (Cat. No. KAI-013), IgG (Cat. No. KAI-014), IgM (Cat. No. KAI-015), and Transferrin (Cat. No. KAI-023) immunoturbidimetric assays. FOR IN VITRO DIAGNOSTIC USE.
Catalog No. NC0609863