Learn More
Multi-Element Plasma Standard Solution 1, Specpure™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042881AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Multi-Element Plasma Standard Solution 1 | |
MFCD00212337 | |
Specpure™ |
100 mL | |
UN3264 | |
Liquid |
Safety and Handling
May be corrosive to metals, Causes severe skin burns and eye damage
Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only