Multi-Element QC-7 Check Standard Solution, Specpure™

Manufacturer: Alfa Aesar 39145AP

Quantity 500mL

 View more versions of this product

Catalog No. AA39145AP

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula 5% HNO3/tr· F?
    MDL Number MFCD00210654
    UN Number UN3264
    Chemical Name or Material Multi-Element QC-7 Check Standard Solution, Standard Solution
    Grade Specpure™

    Specifications

    Quantity 500mL
    TSCA TSCA
    UN Number UN3264
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/show
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Health Hazard 2 Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    Physical Form Liquid
    Appearance Liquid
    MDL Number MFCD00210654
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Molecular Formula 5% HNO3/tr· F?
    Health Hazard 1 H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.
    Chemical Name or Material Multi-Element QC-7 Check Standard Solution, Standard Solution
    Grade Specpure™

    Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/show

    H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit