Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ MULTI GREEN™ ECO Hand Cleaner

GOJO™ MULTI GREEN™ ECO Hand Cleaner is an eco-friendly, multi-purpose hand cleaner with natural pumice scrubbers.

Specifications

Color Green
For Use With (Application) Oil, Grease, Oil Based Paint, Tar, Asphalt, Mud, Clay, Soil, Brake Dust, Carbon Black, Shellacs, Urethanes, Varnish, Wood Stain, Gasket Sealers, Asphalt Sealers, Creosote, Modern Printing Ink
Product Type Heavy Duty Hand Cleaner
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Size Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Size Price Quantity & Availability  
19-043-872
View Documents
GOJO™
235708
8 296mL (10 fl oz.) Each for $6.74
Case of 8 EA for $46.96
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-043-871
View Documents
GOJO™
098904
4 1.89L (0.5 Gallon) Each for $24.65
Case of 4 EA for $86.27
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-043-870
View Documents
GOJO™
235904
4 3.78L (1 Gallon) Each for $36.25
Case of 4 EA for $126.88
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Gel-style hand cleaner clings to hands and spreads easy.
  • Contains natural pumice scrubbers.
  • USDA BioPreferred™ Certified.
  • EcoLogo™ Certified.
  • Readily biodegradeable.
  • Contains a natural citrus solvent.
  • Meets California VOC requirement for consumer products.
  • Effective on oil, grease, paint, tar, and asphalt.
Specifications

Specifications

Green
Oil, Grease, Oil Based Paint, Tar, Asphalt, Mud, Clay, Soil, Brake Dust, Carbon Black, Shellacs, Urethanes, Varnish, Wood Stain, Gasket Sealers, Asphalt Sealers, Creosote, Modern Printing Ink
Heavy Duty Hand Cleaner
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit