Thermo Scientific Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.

Manufacturer: Thermo Scientific 24070290

 View more versions of this product

Catalog No. 14-386-61

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Volume (Metric) Dead <7mL
    Description Tubing set for standard tube dispensing cassette (with 40 cm tubing set
    Autoclavable 50 times at 121°C, 1 bar, 20 min.
    Compatibility Multidrop Combi, Multidrop 384, Multidrop DW
    Diameter Tip: 0.5mm; Tubing: 1.3mm
    Dispensing Range 5 to 2500µL
    Item Description Tubing set for standard tube dispensing
    Material Tip: PP; Tubing: Silicone

    Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.

    Ordering Information:

    Cat Nos 24070297 and 24070290 include tip combs.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit