Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multimedia Package: Elementary Science Products for Science Education

Contains basic elementary slides, transparencies, worksheets and more.

$44.75

Specifications

Category Biology
For Use With (Application) For interactive learning and teaching in school and education
View More Specs

Products 1
Catalog Number Mfr. No. Format Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Format Product Type Includes Price Quantity & Availability  
S06721 Products for Science Education
View Documents
Johannes Lieder GmbH & Co. KG
SSE01
Set Multimedia Student Package: Elementary Science Student package: 6 microscope slides in Plastic box, brochure with explanatory text, cardboard box
Each for $44.75
Add to cart
 
Description

Description

Teacher Package:

  • 6 Microscope slides in plastic box
  • 3 OHP color transparencies
  • 6 Sketch- and worksheets with drawings for all slides
  • Brochure with explanatory text
  • Special cardboard box for storing and packing
Student Set:
  • 6 Microscope slides in plastic box
  • Brochure with explanatory text Cardboard box
Slides:
1. Letter “e” 2. Leg of house fly, w.m. 3. Wing scales of butterfly 4. Human blood smear
5. Large plant cells in the marrow of elderberry, t.s. 6. Colored threads, w.m.

Specifications

Specifications

Biology
For interactive learning and teaching in school and education
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit