Learn More
Multiple-Use Bare Copper Wire
Solid soft drawn annealed copper wire
Manufacturer:  ARCOR ELECTRONIC COMPANY BARE 20GA
 View more versions of this product
Catalog No. 15-545-1C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Copper
QQ-W-343, TYPE S, MIL-W-3861, TYPE S, ASTM-B-3
0.81mm
20
0.25 lb.
Bare Copper Wire
0.03 in.
For busbar crafts, Metal finishers hanging wire and grounding applications
315 Ft./lb.
113g