MultiRAE Plus™ Gas Detection Monitors
Manufacturer: Dawson 0093121014
 View more versions of this product
Catalog No. 19-822-108
  Check Availability
Add to Cart
    Specifications
    Battery Type Lithium ion
    Certifications/Compliance UL, cUL
    Includes Backlit LCD display, built-in two-speed sample pump, rugged rubber boot, external filter and an automatic low flow alarm, vibrating alarm optional, optional datalogging
    For Use With (Application) HazMat/homeland security, confined space entry, environmental, industrial hygiene, plant health and safety
    Product Type Multirae Plus Four-Gas Monitor
    Sensors O2, CO, H2S
  • Description & Specifications
Related Products
Product Content Feedback