Learn More
MP Biomedicals™ Multitest Slide
Features high quality microscope glass with a Teflon™ coating to keep samples in the wells. MP Biomedicals Multitest Slides are resistant to common solvents and are appropriate for one-time use. A 4-channel pipette can be used to transfer samples di
Manufacturer:  MP Biomedicals™ 096040805E
 View more versions of this product
Catalog No. 096040805E
Add to cart
Description
Description
The uncoated areas have a total wettable surface so that liquid spreads evenly over the area of each well. For cell attachment and growth, the slides require only a simple washing in deionized water or detergent followed by sterilization.
  • Self-draining surface
  • Sterilizable
  • Resistant to direct heat
  • Resistant to common organic solvents and fixatives
  • One-time use
  • Numbered
Specifications
Specifications
100 per box
2x4 well, 6mm well diameter