Learn More
MUM1L1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 247621AP
Catalog No. 50-173-4528
Add to cart
Specifications
Specifications
MUM1L1
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5H9M0
MUM1 like protein 1, MUM1L1
Rabbit
IgG
150 μL
-20°C
Primary
139221
Immunohistochemistry (Paraffin)
Unconjugated
MUM1L1
Liquid
MUM1L1
MUM1L1 Fusion Protein Ag20621
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human