Learn More
Mumps virus Mouse anti-Virus, Clone: HM051, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13622
Catalog No. 89-033-585
Add to cart
Description
Description
Mouse monoclonal antibody raised against mumps virus.
  • High specificity
  • High reactivity
Specifications
Specifications
Mumps virus
HM051
Mouse monoclonal antibody raised against mumps virus.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with Nucleoprotein of mumps virus. No cross-reactivity with measles virus, RSV, parainfluenza 1, 2, 3.
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG2b
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)