Learn More
Connecticut Clean Room Corp. Munising LTD Cleanroom Paper
High grade, economical cleanroom paper that contains less synthetic polymers and opacity
$234.00 - $254.00
Specifications
Dimensions (L x W) | 8.5 x 11 in. |
---|---|
Autoclavable | Autoclavable |
For Use With (Application) | Cleanrooms |
Quantity | 2500 Sheets/Cs. |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-807-710
|
Connecticut Clean Room Corp.
PB2S002LTD |
Blue |
Case for $254.00
|
|
|||||
19-807-712
|
Connecticut Clean Room Corp.
PB2S004LTD |
Green |
Case for $254.00
|
|
|||||
19-807-711
|
Connecticut Clean Room Corp.
PB2S003LTD |
Pink |
Case for $254.00
|
|
|||||
19-807-714
|
Connecticut Clean Room Corp.
PB2S001LTD |
White |
Case for $234.00
|
|
|||||
19-807-713
|
Connecticut Clean Room Corp.
PB2S005LTD |
Yellow |
Case for $254.00
|
|
|||||
Description
- Economical alternative to Munising™ LP
- Less synthetic polymers, used to seal individual fibers into sheet
- No natural latex
- Autoclavable
- Free of inorganic fillers, minimizes inadvertent doping of products
- Accepts standard printing inks without skipping, smudging or smearing
- Heat resistant for use in laser printers and photocopy machines
Specifications
8.5 x 11 in. | |
Autoclavable | |
Cleanrooms | |
2500 Sheets/Cs. |