Learn More
Aat BioquestSupplier Diversity Partner MUP [4-Methylumbelliferyl phosphate, free acid] *CAS 3368-04-5*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 11617
A fluorogenic substrate for phosphatases. It is widely used for detecting phosphatases in solution.
Catalog No. 50-195-3371