Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Murashige and Skoog Medium, without Sucrose, Agar, IAA, and Kinetin
Murashige and Skoog Medium is widely used plant tissue culture growth medium. It has proven effective in the culture of tissue derived from moncotyledons and dicotyledons. This medium, originally formulated to support tobacco callus, has been used to support callus initiation and growth, the growth of cells in suspension culture and the regeneration of shoots and plantlets from explants.
Supplier: MP Biomedicals™ 092610029
Description
- Murashige and Skoog Medium contains Murashige and Skoog salts, as well as a mixture of vitamins.
- This modification of the original formula does not contain agar, sucrose, IAA, and Kinetin.
- Murashige and Skoog Medium is widely used plant tissue culture growth medium.
- It has proven effective in the culture of tissue derived from moncotyledons and dicotyledons.
- This medium, originally formulated to support tobacco callus, has been used to support callus initiation and growth, the growth of cells in suspension culture and the regeneration of shoots and plantlets from explants.
- More recently, it has been used for micropropagation of ornamental, vegetable and fruit plants in research and commercial enterprises.
Specifications
Store at +4°C. | |
Cell Culture Media | |
Component: Amino Acids-Glycine: 2.00000mg/L; Vitamins- myo-Inositol: 100.00000mg/L; Niacin: 0.50000mg/L; Pyridoxine HCl: 0.50000mg/L; Thiamine HCl 0.10000mg/L; Inorganic Salts-Ammonium Nitrate [NH4NO3]: 1650.00000mg/L; Boric Acid [H3BO3]: 6.20000mg/L; Calcium Chloride [CaCl2 2H2O] (anhyd): 332.0000mg/L; Cobalt Chloride [CoCl2 6H2O] Hexahydrate: 0.02500mg/L; Cupric Sulfate [CuSO4]: 0.01600mg/L; EDTA Fe Na: 36.70000mg/L; Magnesium Sulfate [MgSO4]: 180.70000mg/L; Manganese Sulfate [MnSO4 H2O] Monohydrate: 16.90000mg/L; Potassium Iodide [KI]: 0.83000mg/L; Potassium Nitrate [KNO3]: 1900.00000mg/L; Potassium Phosphate Monobasic [KH2PO4]: 170.00000mg/L; Sodium Molybdate [Na2MoO4 2H2O] Dihydrate: 0.25000mg/L; Zinc Sulfate [ZnSO4 7H2O] Heptahydrate: 8.60000mg/L | |
4.80 to 5.65(before additions) |
Powder | |
<0.5 EU/mL | |
Plant, Tissue Culture | |
1 x 100 L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction