Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murexide, 0.25% w/w in Sucrose, Reagents

Catalog No. 01334770
Change view
Click to view available options
Quantity:
100 mL
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-770 250 g
01-334-771 100 mL
01-334-769 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-770 Supplier Reagents Holdings Llc Supplier No. CM040000250C2
Only null left
Add to Cart
Add to Cart

Murexide, 0.25% w/w in Sucrose, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 57-50-1 , 3051-09-0
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym Ammonium Purpurate, Color Index 56085
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Percent Purity 0.25%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.587
Color Pink
Quantity 250 g
Physical Form Solid
Chemical Name or Material Murexide, 0.25% w/w in Sucrose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.