Learn More
MURF1 Monoclonal Antibody (HL1450), Invitrogen™
Rabbit Monoclonal Antibody
Supplier:  Thermo Scientific MA547051
Encompass
Catalog No. PIMA547051
Add to Cart
Description
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
MURF1 Monoclonal antibody specifically detects MURF1 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MURF1
Monoclonal
1 mg/mL
PBS with no preservative
Q38HM4, Q969Q1
TRIM63
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human MURF1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot
HL1450
Unconjugated
TRIM63
E3 ubiquitin-protein ligase TRIM63; FLJ32380; IRF; iris ring finger protein; Murf; MURF 1; MuRF1; MuRF-1; MURF2; muscle ring finger protein 1; muscle specific ring finger protein 2; muscle-specific RING finger protein 1; OTTHUMP00000008701; RF1; ring finger protein 28; RING-type E3 ubiquitin transferase TRIM63; RNF 28; RNF28; SMRZ; striated muscle RING zinc finger protein; TRIM 63; Trim63; tripartite motif containing 63; tripartite motif containing 63, E3 ubiquitin protein ligase; tripartite motif protein 63; tripartite motif-containing 63; tripartite motif-containing protein 63
Rabbit
Protein A
RUO
433766, 84676
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations
Recommendations