Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Musashi-2 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3255R100UG
This item is not returnable.
View return policy
Description
Musashi-2 Monoclonal specifically detects Musashi-2 in Human samples. It is validated for Flow Cytometry.Specifications
Musashi-2 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
FLJ36569, MGC3245, MSI2, MSI2H, musashi homolog 2 (Drosophila), musashi-2, Musashi2, RNA-binding protein Musashi homolog 2 | |
E. coli-derived recombinant human Musashi-2 Met1-His328 Accession # Q96DH6 | |
RUO | |
124540 | |
Human | |
IgG2b |
Flow Cytometry | |
960121 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q96DH6 | |
Mouse | |
100 μg | |
Primary | |
Detects human Musashi-2 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction