Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Musashi-2 Rabbit anti-Human, Clone: RM422, Novus Biologicals™

Rabbit Monoclonal Antibody

$104.00 - $366.50

Specifications

Antigen Musashi-2
Clone RM422
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:400
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB119698
View Documents
Novus Biologicals
NBP30917525UL
25 μg
Each for $104.00
Add to cart
 
NB119697
View Documents
Novus Biologicals
NBP309175100UL
100 μg
Each for $366.50
Add to cart
 
Description

Description

Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Musashi-2
Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:400
Unconjugated
Rabbit
Neuronal Cell Markers
PBS, 50% Glycerol, 1% BSA
124540
IgG
Primary
Store at -20°C.
RM422
Western Blot, Immunohistochemistry
Purified
RUO
Human
FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2
A peptide corresponding to the internal region of human Musashi-2
Monoclonal
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit