Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Muscarinic Acetylcholine Receptor M3/CHRM3 Antibody (580011), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6378MFV450
Description
Muscarinic Acetylcholine Receptor M3/CHRM3 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M3/CHRM3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| Muscarinic Acetylcholine Receptor M3/CHRM3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 1131 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 580011 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| cholinergic receptor, muscarinic 3, HM3, m3 muscarinic receptor, muscarinic acetylcholine receptor M3 | |
| NS0 mouse myeloma cell line transfected with human CHRM3, Accession # P20309 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction