Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Muscarinic Acetylcholine Receptor M5/CHRM5 Antibody (590123), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10323PEATT594
Description
Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for Flow CytometrySpecifications
Muscarinic Acetylcholine Receptor M5/CHRM5 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
590123 | |
Flow Cytometry | |
cholinergic receptor, muscarinic 5, HM5, MGC41838, muscarinic acetylcholine receptor M5 | |
NS0 mouse myeloma cell line transfected with human CHRM5, Met1-Pro532, Accession # P08912 | |
0.1 mL | |
GPCR, Neuroscience | |
1133 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction