Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscle Phosphofructokinase/PFKM/PFK-1 Rabbit anti-Human, Mouse, Alexa Fluor™ 532, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP232169AF532

Catalog No. NP232169A32


Add to cart

Description

Description

Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Muscle Phosphofructokinase/PFKM/PFK-1
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Mouse
Western Blot
Western Blot
Affinity Purified, Conjugated
PFKM
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Phosphofructokinase, Muscle using the numbering given in entry NP_000280.1 (GeneID 5213).
Immunogen affinity purified
Lipid and Metabolism
Primary
5213.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit