Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Rabbit anti-Human, Mouse, Alexa Fluor™ 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232169AF594
Catalog No. NP232169A94
Add to cart
Description
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human,Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Muscle Phosphofructokinase/PFKM/PFK-1
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
PFKM
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Phosphofructokinase, Muscle using the numbering given in entry NP_000280.1 (GeneID 5213).
100 μL
Primary
5213
Western Blot
Western Blot
6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse