Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 170341AP150UL
Description
MUT encodes the mitochondrial enzyme methylmalonyl Coenzyme A mutase. In humans, the product of this gene is a vitamin B12-dependent enzyme which catalyzes the isomerization of methylmalonyl-CoA to succinyl-CoA, while in other species this enzyme may have different functions. Mutations in this gene may lead to various types of methylmalonic aciduria.Specifications
MUT | |
Polyclonal | |
Unconjugated | |
MUT | |
MCM, Methylmalonyl CoA isomerase, MUT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17850, 4594, 688517 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16332, P22033 | |
Mut | |
MUT Fusion Protein Ag10523 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction