Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUT Mouse anti-Human, Clone: OTI2C8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526287

Catalog No. PIMA526287


Add to cart

Description

Description

This gene encodes the mitochondrial enzyme methylmalonyl Coenzyme A mutase. In humans, the product of this gene is a vitamin B12-dependent enzyme which catalyzes the isomerization of methylmalonyl-CoA to succinyl-CoA, while in other species this enzyme may have different functions. Mutations in this gene may lead to various types of methylmalonic aciduria.
Specifications

Specifications

MUT
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P22033
MUT, MCM
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4594
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2C8
Unconjugated
MUT
Liquid
MUT
Full length human recombit protein of MUT produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit