Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mutant HTT Mouse anti-Human,Mouse, Unconjugated, Clone: PHP1, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen mutant HTT
Applications Dot Blot, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN24901MI
View Documents
MilliporeSigma
MABN2490100UL
100 μL
Each for $372.00
Add to cart
 
MABN24902MI
View Documents
MilliporeSigma
MABN249025UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

mutant HTT Monoclonal antibody specifically detects mutant HTT in Human,Mouse samples. It is validated for Dot Blot, Electron Microscopy, Immunohistochemistry (Paraffin), Inhibits Activity/Function, Western Blotting.
Specifications

Specifications

mutant HTT
Unconjugated
Mouse
Huntingtin;Huntington disease protein;HD protein
GST-tagged recombinant exon1 of wild-type human huntingtin HTTx1 (20Q) protein.
Primary
Stable for 1 year at -20°C from date of receipt.
Dot Blot, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse
HTT, HD, IT15
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit