Learn More
Aviva Systems Biology Corp MVP Antibody - N-terminal region (ARP58642_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58642_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MVP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MVP. Alternate names: LRP, VAULT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62385