Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVP Mouse anti-Human, Clone: 1032, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14710

Catalog No. 89-034-793


Add to cart

Description

Description

Mouse monoclonal antibody raised against human MVP.

  • High specificity
  • High reactivity
Specifications

Specifications

MVP
1032
Mouse monoclonal antibody raised against human MVP.
In 10 mM PBS.
Liquid
MVP
Proteins precipitated from human breast cancer MCF-7 cell line.
Protein A/G purification
RUO
Primary
9961
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q14764
LRP|VAULT1
Mouse
IgG, kappa
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Recognizes a protein of 104kDa-110kDa which characterized as MVP.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)