Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVP Rabbit anti-Human, Clone: VP2897R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen MVP
Clone VP2897R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122433
View Documents
Novus Biologicals
NBP30748010UG
100 μg
Each for $406.52
Add to cart
 
NB122434
View Documents
Novus Biologicals
NBP30748020UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

MVP Monoclonal antibody specifically detects MVP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MVP
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
LRPVAULT1, Lung resistance-related protein, major vault protein
Recombinant full-length human MVP protein
Monoclonal
Protein A or G purified
VP2897R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Signal Transduction
10 mM PBS with 0.05% BSA
9961
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit