Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

MxA/Mx1 Mouse anti-Human, HRP, Clone: 683, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243786H

Catalog No. NBP243786H



Description

Description

MxA/Mx1 Monoclonal antibody specifically detects MxA/Mx1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

MxA/Mx1
683
Western Blot
Purified
MX1
Recombinant protein encompassing a sequence within the center region of human MX1. The exact sequence is proprietary.
Protein G purified
Apoptosis
Primary
4599
Western Blot
HRP
PBS with No Preservative
human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse)
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit