Learn More
MXI1 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29927
Catalog No. 89-033-215
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human MXI1.
  • High specificity
  • High reactivity
Specifications
Specifications
MXI1
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user.
NM_130439
MAD2|MGC43220|MXD2|MXI|bHLHc11
Rabbit
Affinity purification
RUO
Primary
4601
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human MXI1.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
MXI1
A synthetic peptide corresponding to internal region of human MXI1.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)