Learn More
Tocris Bioscience™ MY-5445
PDE5 inhibitor
Supplier: Tocris Bioscience™ 0432/50
This item is not returnable.
View return policy
Description
Specific inhibitor of cyclic GMP phosphodiesterase, selective for PDE5 (IC50 = 0.5 μM).Specifications
1-(3-Chlorophenylamino)-4-phenylphthalazine | |
50 mg | |
C20H14ClN3 |
78351-75-4 | |
Soluble to 50 mM in DMSO and to 5 mM in ethanol | |
Observed MW: 331.8 |
Safety and Handling
Recommended Storage : Store at Room Temperature