Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYADM Mouse anti-Human, Clone: MYADM/972, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14421

Catalog No. 89-034-504


Add to cart

Description

Description

Mouse monoclonal antibody raised against human MYADM.

  • High specificity
  • High reactivity
Specifications

Specifications

MYADM
MYADM/972
Mouse monoclonal antibody raised against human MYADM.
In 10 mM PBS (0.05% BSA and 0.05% azide).
Liquid
MYADM
Recombinant protein corresponding to human MYADM.
Protein A/G purification
RUO
Primary
91663
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q96S97
SB135
Mouse
IgG1
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)