Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myb Monoclonal Antibody (IOA3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA190154
Description
Predicted molecular weight of ~77 kDa.
Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3'. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.
Specifications
Myb | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P06876, P10242 | |
MYB | |
Recombinant v-myb protein, DNA binding protein. | |
100 μg | |
Primary | |
Human, Mouse, Frog, Zebrafish | |
Antibody | |
IgG2a |
Immunoprecipitation, Western Blot | |
IOA3 | |
Unconjugated | |
MYB | |
AI550390; Cmyb; c-myb; cmyb protein; c-myb_CDS; c-myb10A_CDS; c-myb13A_CDS; c-myb14A_CDS; c-myb8B_CDS; efg; gag-myb protein; M16449; MYB; myb proto-oncogene protein; MYB proto-oncogene, transcription factor; Mybl2; myb-related protein B; myeloblastosis oncogene; myeloblastosis oncogene-like 2; myeloblastosis proto-oncogene product; oncogene AMV; proto-oncogene c-Myb; RP1-32B1.1; transcription factor cmyb; transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog | |
Mouse | |
Protein G | |
RUO | |
17863, 30519, 4602 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction