Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

v-Myb Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548879

Catalog No. PA548879

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Strong transcriptional activator; DNA-binding protein that specifically recognize the sequence 5 ft.-YAAC[GT]G-3 ft.. Could have a role in the proliferation and/or differentiation of neurogenic, spermatogenic and B-lymphoid cells.
Specifications

Specifications

v-Myb
Polyclonal
PBS with 0.09% sodium azide
P10243
Myb-related protein A, A-Myb, Myb-like protein 1, MYBL1, AMYB
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4603
Western Blot
Unconjugated
MYBL1
Liquid
MYBL1
KLH conjugated synthetic peptide between 510-536 amino acids from the Central region of human MYBL1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit