Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH6 Mouse anti-Human, Clone: CL2148, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524631

Catalog No. PIMA524631


Add to cart

Description

Description

Immunogen sequence: QVEEDKVNSL SKSKVKLEQQ VDDLEGSLEQ EKKVRMDLER AKRKLEGDLK LTQESIMDLE NDKLQLEEKL KKKEFDINQQ NSKIEDEQVL ALQLQKKLKE NQARIEELEE ELEAERTARA KVEKLRSDLS RELEEISERL EEA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%. Binds to an epitope located within the peptide sequence LTQESIMDLENDKLQ as determined by overlapping synthetic peptides.

3-48 antibody cross-reacts with heavy chains of human myosin isolated from: slow skeletal muscle, fast skeletal muscle, eye muscle, and streptococcal M5 peptide. It does not react with human platelet, smooth muscle myosin heavy chains, myosin light chains, myosin fragments circulating in blood of patients with myocardial infarction, nor does it react with any other human tissue antigens or blood cells.
Specifications

Specifications

MYH6
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P13533
MYBPC, MYBPCF; C-protein, skeletal muscle fast isoform; fast MyBP-C; fast-type muscle myosin-binding-protein C; myosin-binding protein C, fast-type
Mouse
IgG1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4624
Immunohistochemistry, Western Blot
CL2148
Unconjugated
MYH6
Liquid
MYH6
Recombit protein corresponding to Human MYBPC2
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only