Learn More
MYBPH Mouse anti-Human, Clone: OTI3G1, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA505664
Catalog No. 50-167-6096
Add to cart
Description
Description
MYBPH binds to myosin; probably involved in interaction with thick myofilaments in the A-band.
Specifications
Specifications
MYBPH
Monoclonal
Unconjugated
MYBPH
Liquid
MYBPH
Full length human recombit protein of human MYBPH produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3G1
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13203
myosin binding protein H
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4608