Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYC, anti-Human, Clone: 9E10, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00163

Catalog No. 89-032-506


Add to cart

Specifications

Specifications

MYC
9E10
Immunofluorescence Immunohistochemistry (Frozen sections) (5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
v-myc myelocytomatosis viral oncogene homolog (avian)
bHLHe39|c-Myc
Original antibody is raised against a synthetic peptide corresponding to amino acids 408-439 of human c-myc.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
4609
Human
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
MYC
IgG2a, kappa
RUO
Antibody
Monoclonal
The epitope sequence, EQKLISEEDL, is located in aa 410-419 of human c-myc protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit