Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYC, anti-Human, Clone: 9E10, Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00175
Specifications
MYC | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
bHLHe39/c-Myc | |
Original antibody is raised against a synthetic peptide corresponding to amino acids 408-439 of human c-myc. | |
RUO | |
4609 | |
Human | |
Antibody | |
IgG2b κ |
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
9E10 | |
Immunofluorescence Immunohistochemistry (Frozen sections) (5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
v-myc myelocytomatosis viral oncogene homolog (avian) | |
MYC | |
200 μg | |
Primary | |
The epitope sequence, EQKLISEEDL, is located in aa 410-419 of human c-myc protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction