Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc Epitope Tag, Armenian Hamster anti-Human, Clone: 90000000000, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen Myc Epitope Tag
Clone 90000000000
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Host Species Armenian Hamster
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB014113
View Documents
Novus Biologicals
NBP2810190.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB014114
View Documents
Novus Biologicals
NBP2810190.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Myc Epitope Tag Monoclonal antibody specifically detects Myc Epitope Tag in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry-Frozen.
Specifications

Specifications

Myc Epitope Tag
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Human
Primary
90000000000
Armenian Hamster
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit