Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc Epitope Tag Mouse anti-Human, Alexa Fluor™ 750, Clone: 2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243627AF750

Catalog No. NP243627A70


Add to cart

Description

Description

Myc Epitope Tag Monoclonal antibody specifically detects Myc Epitope Tag in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Myc Epitope Tag
2
Western Blot, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
The exact sequence is proprietary.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1
0.1 mL
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit