Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYC tag Goat anti-Tag, Polyclonal, liquid, TrueMAB™

Goat Polyclonal Antibody
Supplier: OriGene TA150083

Description
Synthetic peptide sequence is EQKLISEEDL
Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.Specifications
MYC tag | |
Polyclonal | |
Whole serum with 0.05% sodium azide | |
Goat | |
150 μL | |
Primary | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
anti-Myc, tag antibody, Anti-myc Antibody, anti-myc antibodies, myc antibody, cmyc antibody, c-myc antibody, anti myc, anti cmyc, anti c-myc, 9E10 myc, 9E10 anti myc, 9E10 antibody | |
Synthetic peptide conjugated to KLH | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction