Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc-Tag Mouse anti-Human, Active Motif™ Non-distribution product as customer accommodation.

Mouse Recombinant Monoclonal Antibody

$98.40 - $465.85

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen Myc-Tag
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3740 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91204
10 μg
Each for $98.40
Add to cart
 
50-199-3739 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91203
100 μg
Each for $465.85
Add to cart
 
Description

Description

Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.
Specifications

Specifications

Monoclonal
Primary
Myc-Tag
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit