Learn More
Mycobacterium smegmatis Lactate oxidase Rabbit anti-Bacteria, Polyclonal, Nordic

Rabbit Polyclonal Antibody
Supplier: Vector Laboratories NE083/7S

Description
Prior to use, an aliquot is thawed slowly at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used t he same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the product.
Mycobacterium smegmatis Lactate oxidase Polyclonal antibody specifically detects Mycobacterium smegmatis Lactate oxidase in Bacteria samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry, ImmunofluorescenceSpecifications
Mycobacterium smegmatis Lactate oxidase | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence | |
10 mg/mL | |
PBS with no preservative; pH | |
Mycobacterium smegmatis Lactate oxidase | |
10 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.