Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium Tuberculosis 16kDa Mouse anti-Bacteria, DyLight 488, Clone: HTM63, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB11018070G

Catalog No. NB11018070G

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mycobacterium Tuberculosis 16kDa Monoclonal antibody specifically detects Antigen in Bacteria samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Mycobacterium Tuberculosis 16kDa
HTM63
Western Blot, ELISA
Purified
Recombinant full length protein: M. tuberculosis 16 kDa antigen.
Protein G purified
RUO
Primary
recognises native and recombinant M. tuberculosis 16 kDa antigen.
Western Blot, ELISA
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Bacteria
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit