Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium tuberculosis Ag85b Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5122320

This item is not returnable.
View return policy
Description
Mycobacterium tuberculosis Ag85b Polyclonal antibody specifically detects Mycobacterium tuberculosis Ag85b in Bacteria samples. It is validated for ELISA, Western BlotSpecifications
Mycobacterium tuberculosis Ag85b | |
Polyclonal | |
Unconjugated | |
ag85B | |
Recombinant Mycobacterium tuberculosis Ag85b produced in E. coli | |
100 μg | |
Primary | |
Bacteria | |
Antibody |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23491943 | |
4° C, do not freeze | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title