Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium Tuberculosis ESAT6 Mouse, Clone: RSB14, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Mycobacterium Tuberculosis ESAT6 |
---|---|
Clone | RSB14 |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105468SS
|
Novus Biologicals
NBP105468SS |
0.05 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NBP105468
|
Novus Biologicals
NBP105468 |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Mycobacterium Tuberculosis ESAT6 Monoclonal antibody specifically detects Antigen in Non-species specific samples. It is validated for Enzyme Linked Immunosorbent Assay.Specifications
Mycobacterium Tuberculosis ESAT6 | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
6 kDa early secretory antigenic target, 6 kDa early secretory antigenic target ESXA, ESAT 6, esaT6, ESXA | |
IgG1 | |
Protein G purified |
RSB14 | |
ELISA | |
Unconjugated | |
Mouse | |
Nonspecies specific | |
ESAT antigen. | |
Primary | |
Store at 4C. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title