Learn More
Selleck Chemical LLCSupplier Diversity Partner Mycophenolate Mofetil 100mg 128794-94-5 RS61443  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1501100MG
Mycophenolate Mofetil (RS61443) is a non-competitive, selective and reversible inhibitor of inosine monophosphate dehydrogenase I/II with IC50 of 39 nM and 27 nM, respectively. Mycophenolate Mofetil induces caspase-dependent apoptosis and cell cycle inhibition in multiple myeloma cells.
Catalog No. 50-635-1