Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycoplasma bovis Mouse anti-Bacteria, Alexa Fluor™ 532, Clone: 10F181, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP230167AF532

Catalog No. NP230167A32


Add to cart

Description

Description

Mycoplasma bovis Monoclonal antibody specifically detects Mycoplasma bovis in Bacteria samples. It is validated for ELISA.
Specifications

Specifications

Mycoplasma bovis
10F181
ELISA
Conjugated, Purified
M. bovis preparation
Protein A purified
Store at 4°C in the dark.
Monoclonal
Bacteria
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1
0.1 mL
Primary
Recognizes Mycoplasma bovis. Does not react with Streptococcus agalactaie, S. aureus, E. coli, M. bovigentalium or M. californcum.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit